
Potassium bromate 1/60 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur.
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7758-01-2
:
Formula:KBrO₃
MDL Number:MFCD00011359
:167 g/mol
EINECS:231-829-8
:GHS08
Storage Temperature:Ambient
Frequently Bought Together