
Orthophosphoric acid 50% analytical reagent
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7664-38-2
:
Formula:H₃PO₄
MDL Number:MFCD00011340
:98 g/mol
Density:1.5…1.8 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
:GHS05
Frequently Bought Together