Sodium hydrogen carbonate 0.5 mol/l in aqueous solution
Catalog # ACSDEL002.W.L5
Supplier: CPACHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
Specifications
About this item
Synonyms: Sodium bicarbonate
:
:84.01 g/mol
Storage Temperature:Ambient
EINECS:205-633-8
CAS Number:144-55-8
Formula:NaHCO₃
MDL Number:MFCD00003528