Sodium carbonate 20% in aqueous solution analytical reagent
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:497-19-8
:
Formula:Na₂CO₃
MDL Number:MFCD00003494
:105.99 g/mol
Storage Temperature:Ambient
EINECS:207-838-8
Frequently Bought Together