Order Entry
Belgium
ContactUsLinkComponent
Bismuth standard solution, 1,000 mg/L Bi in 10% nitric acid (from Bi) standard for AAS and ICP
Bismuth standard solution, 1,000 mg/L Bi in 10% nitric acid (from Bi) standard for AAS and ICP
Catalog # PERCN9300105
Supplier:  PerkinElmer
CAS Number:  
Bismuth standard solution, 1,000 mg/L Bi in 10% nitric acid (from Bi) standard for AAS and ICP
Catalog # PERCN9300105
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    10% HNO₃
  • Pk:
    1
  • Element:
    Bismuth
  • Symbol:
    Bi

Specifications

About this item