Order Entry
Belgium
ContactUsLinkComponent
J.T.Baker®, Bismuth Standard Solution, 1000 mg/L Bi in dil. Nitric Acid (from Bi) Standard for AAS
J.T.Baker®, Bismuth Standard Solution, 1000 mg/L Bi in dil. Nitric Acid (from Bi) Standard for AAS
Catalog # 6922.0100
Supplier:  Avantor
CAS Number:  
J.T.Baker®, Bismuth Standard Solution, 1000 mg/L Bi in dil. Nitric Acid (from Bi) Standard for AAS
Catalog # 6922.0100
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Bismuth
  • Symbol:
    Bi

Specifications

About this item

Certifications: Traceable to NIST