Bismuth standard solution, 1,000 mg/L Bi in dil. nitric acid standard for ICP
Catalog # ACCUICP06N5
Supplier: AccuStandard
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Glass bottle
- Pk:500 mL
- Element:Bismuth
- Symbol:Bi