Order Entry
Belgium
ContactUsLinkComponent
J.T.Baker®, Beryllium Standard Solution, 1000 mg/L Be in dil. Nitric Acid (from BeO.(Be(OAc)₂)₃) Standard for ICP
J.T.Baker®, Beryllium Standard Solution, 1000 mg/L Be in dil. Nitric Acid (from BeO.(Be(OAc)₂)₃) Standard for ICP
Catalog # 5706.0100
Supplier:  Avantor
CAS Number:  
J.T.Baker®, Beryllium Standard Solution, 1000 mg/L Be in dil. Nitric Acid (from BeO.(Be(OAc)₂)₃) Standard for ICP
Catalog # 5706.0100
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    01
  • Environmentally Preferable:
  • Element:
    Beryllium
  • Symbol:
    Be

Specifications

About this item

Certifications: Traceable to NIST.