Order Entry
Belgium
Orders LinkContactUsLinkComponent
Hydrochloric acid 3 mol/l (3 N)

Hydrochloric acid 3 mol/l (3 N)

Glass bottle, 1 L
Catalog # REGEH23001
Irritant
Supplier:   REAGECON
Hydrochloric acid 3 mol/l (3 N)
Catalog # REGEH23001
Supplier:  REAGECON
CAS Number:  7647-01-0
Hydrochloric acid 3 mol/l (3 N)

Glass bottle, 1 L

CAS Number:  7647-01-0
Price
68,40 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Hydrochloric acid solution, Muriatic acid
  • CAS Number:7647-01-0
  • Formula:HCl
  • MDL Number:MFCD00011324
  • Molecular Weight:36.46 g/mol
  • Density:1.06 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-595-7
  • UN:1789
  • ADR:8,II

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications