Order Entry
Belgium
ContactUsLinkComponent
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis), granular USP, J.T.Baker®
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis), granular USP, J.T.Baker®
Catalog # C4908-88
Supplier:  Avantor
CAS Number:  
undefined
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis), granular USP, J.T.Baker®
Catalog # C4908-88
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • GMP Manufactured Product:
    Passes Test
  • Meets U.S.P Requirements:
    Passes Test
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤1.0%
  • Bulk Pharmaceutical Chemical:
    97.5 - 102.0%
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Identification C: Water (H₂O):
    ≤1.0 %
  • USP - Assay (Anhydrous basis):
    97.5 - 102.0 %
  • USP - Related substances: Maltose and isomaltose:
    ≤0.4 %
  • USP - Related substances: Maltotriose:
    ≤0.2 %
  • USP - Related substances: Fructose:
    ≤0.15 %
  • USP - Related substances: Unspecified impurities, each:
    ≤0.10 %
  • USP - Related substances: Total Impurities:
    ≤0.5 %
  • USP - Color and Clarity of Solution:
    Passes Test
  • USP - Soluble Starch, Sulfites:
    ≤15 ppm
  • USP - Conductivity (uS cm-1):
    ≤20 µS/cm
  • USP - Dextrin:
    Passes Test
  • USP - Endotoxin Concentration (EU/g):
    ≤2.5
  • Solubility:
    Passes Test

Specification Test Results

About this item

 :
Merck Index:14,04459
Melting Pt:146 °C
Density:1.54 g/cm³ (20 °C)
 :180.16 g/mol
Storage Temperature:Ambient
EINECS:200-075-1
CAS Number:50-99-7
 :492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912