Order Entry
Belgium
ContactUsLinkComponent
 
Lead standard solution, 1000 mg/L Pb in dil. nitric acid (from Pb(NO₃)₂) TraceCERT® standard for ICP, Supelco®
  SUPL41318-100ML-F
 :  Merck
 :  
Lead standard solution, 1000 mg/L Pb in dil. nitric acid (from Pb(NO₃)₂) TraceCERT® standard for ICP, Supelco®
  SUPL41318-100ML-F
 :  Merck
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Lead
  • Symbol:
    Pb

 

 

 : Traceable to SRM from NIST, tested in ISO 17034 /ISO 17025 accredited laboratory.