Order Entry
Belgium
ContactUsLinkComponent
J.T.Baker® BAKER INSTRA-ANALYZED®, Lead Standard Solution, 1000 mg/L Pb in 5% Nitric Acid (from Pb) Standard for ICP
J.T.Baker® BAKER INSTRA-ANALYZED®, Lead Standard Solution, 1000 mg/L Pb in 5% Nitric Acid (from Pb) Standard for ICP
Catalog # 5765.0100
Supplier:  Avantor
CAS Number:  
J.T.Baker® BAKER INSTRA-ANALYZED®, Lead Standard Solution, 1000 mg/L Pb in 5% Nitric Acid (from Pb) Standard for ICP
Catalog # 5765.0100
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    02
  • Element:
    Lead
  • Symbol:
    Pb

Specifications

About this item

Certifications: Traceable to NIST.