Aluminium standard solution, 10000 mg/L Al in 2 M nitric acid (from Al) standard for AAS
Catalog # KRAF12847.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:2 M HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Aluminium
- Symbol:Al