Order Entry
Belgium
ContactUsLinkComponent
J.T.Baker®, Barium Standard Solution, 1000 mg/L Ba in dil. Nitric Acid (from Ba(NO₃)₂) Standard for AAS
J.T.Baker®, Barium Standard Solution, 1000 mg/L Ba in dil. Nitric Acid (from Ba(NO₃)₂) Standard for AAS
Catalog # 6920.0100
Supplier:  Avantor
CAS Number:  
J.T.Baker®, Barium Standard Solution, 1000 mg/L Ba in dil. Nitric Acid (from Ba(NO₃)₂) Standard for AAS
Catalog # 6920.0100
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pk:
    1
  • Element:
    Barium
  • Symbol:
    Ba

Specifications

About this item

Certifications: Traceable to NIST.