Order Entry
Belgium
ContactUsLinkComponent
Barium standard solution, 1,000 mg/L Ba in dil. nitric acid standard for ICP-MS
Barium standard solution, 1,000 mg/L Ba in dil. nitric acid standard for ICP-MS
Catalog # ACCUICPMS04N0.1X1
Supplier:  AccuStandard
CAS Number:  
Barium standard solution, 1,000 mg/L Ba in dil. nitric acid standard for ICP-MS
Catalog # ACCUICPMS04N0.1X1
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Barium
  • Symbol:
    Ba

Specifications

About this item