Order Entry
Belgium
ContactUsLinkComponent
Aluminium standard solution, 1000 mg/L Al in 0,5 M nitric acid standard for AAS
Aluminium standard solution, 1000 mg/L Al in 0,5 M nitric acid standard for AAS
Catalog # ACRO195805000
CAS Number:  
Aluminium standard solution, 1000 mg/L Al in 0,5 M nitric acid standard for AAS
Catalog # ACRO195805000
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Aluminium
  • Symbol:
    Al

Specifications

About this item