

2-Aminophenol
Catalog # PROOMM0217.06
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
Merck Index:13,00460
Boiling Pt:235 °C (1013 hPa)
Density:1.328 g/cm³ (20 °C)
Flash Pt:>175 °C
EINECS:202-431-1
UN:2512
ADR:6.1,III
:109.13 g/mol
:GHS08
:GHS07
CAS Number:95-55-6
Formula:C₆H₇NO
MDL Number:MFCD00007690