Order Entry
Belgium
ContactUsLinkComponent
VWR® Traceable® Ultra, Digital Dial Thermometers
VWR® Traceable® Ultra, Digital Dial Thermometers
Catalog # 620-1665
Supplier:  VWR Collection
undefined
VWR® Traceable® Ultra, Digital Dial Thermometers
Catalog # 620-1665
Supplier:  VWR Collection

Specifications

  • Ø:
    38 mm
  • Range:
    –50…+300 °C
  • Resolution:
    0,1 °C
  • Weight:
    32 g
  • Accuracy:
    ±0,4 °C
  • Pk:
    1

Specifications

About this item

Dial thermometer for use in soil, cuvettes, test tubes, food products, incubators, biomedical reagents and petroleum hydrocarbons.

  • Easy to read
  • Large, bright LCD

Stainless steel probe (Lר: 120×4,8 mm).

Certifications: Calibration complies with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Delivery: Supplied with protective case, silver-oxide battery and certificate of calibration.