Orders LinkContactUsLinkComponent
Adapters, glass, cone/screwthread
Adapters, glass, cone/screwthread
Supplier:   Lenz Laborglas
Adapters, glass, cone/screwthread
Supplier:  Lenz Laborglas
Certificates
Adapters, glass, cone/screwthread
Adapters, glass, cone/screwthread
11 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread adapters, with screw-cap with aperture and silicone sealing ring with PTFE washers.
Documents
Product Family Options
Product Information
  • Cone size
    Thread
    Tube ext.Ø
    Availability
    Price
  • 14/23 NS
    GL 14
    5,5 - 6,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,80 €
    16,80 €
    1 * 1 ST
       16,80 €
  • 19/26 NS
    GL 14
    5,5 - 6,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,10 €
    19,10 €
    1 * 1 ST
       19,10 €
  • 24/29 NS
    GL 14
    5,5 - 6,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,10 €
    29,10 €
    1 * 1 ST
       29,10 €
  • 29/32 NS
    GL 14
    5,5 - 6,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,30 €
    29,30 €
    1 * 1 ST
       29,30 €
  • 14/23 NS
    GL 18
    7,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,50 €
    21,50 €
    1 * 1 ST
       21,50 €
  • 19/26 NS
    GL 18
    7,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,30 €
    25,30 €
    1 * 1 ST
       25,30 €
  • 29/32 NS
    GL 18
    7,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,90 €
    27,90 €
    1 * 1 ST
       27,90 €
  • 19/26 NS
    GL 25
    9 - 11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,80 €
    24,80 €
    1 * 1 ST
       24,80 €
  • 24/29 NS
    GL 25
    9 - 11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,60 €
    24,60 €
    1 * 1 ST
       24,60 €
  • 29/32 NS
    GL 25
    9 - 11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,10 €
    36,10 €
    1 * 1 ST
       36,10 €
  • 29/32 NS
    GL 32
    13 - 15 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,80 €
    42,80 €
    1 * 1 ST
       42,80 €