Micro-fluidic platform, SilFlow™
About this item
SilFlow™ microchannel device (MCD) options for three-port GC splitters, four-port GC splitters and a Deans' switch MCD, perfect for multi-dimensional analyses. SilFlow™ can be incorporated into the system without impacting the chromatography.
- Chemically-deactivated stainless steel channels avoiding active sites experienced with conventional connections
- Conventional graphite or Vespel® ferrules cannot be used with the SilFlow™ MCD as the dimensional stability is not adequate and there is a risk of contaminating the channels
- SilTite™ FingerTite metal ferrules result in a reliable zero dead volume connection, giving optimised peak shapes
- Can be operated at pressures greater than 25000 psi
- SilFlow™ tracks oven temperature up to 20 °C/min. The design of SilFlow™ alleviates cold spots and sample condensation
: SilFlow™ kits contain: SilFlow™ microchannel device (MCD), SilTite™ FingerTite tool, mounting bracket, appropriately sized ferrules, nuts and blanking ferrules for set up. 1,1 mm Ø tubing is not supplied in the splitter kits and must be ordered separately.
30 Options Available Below
Product Details & Documents
SilFlow™ microchannel device (MCD) options for three-port GC splitters, four-port GC splitters and a Deans' switch MCD, perfect for multi-dimensional analyses. SilFlow™ can be incorporated into the system without impacting the chromatography.
- Chemically-deactivated stainless steel channels avoiding active sites experienced with conventional connections
- Conventional graphite or Vespel® ferrules cannot be used with the SilFlow™ MCD as the dimensional stability is not adequate and there is a risk of contaminating the channels
- SilTite™ FingerTite metal ferrules result in a reliable zero dead volume connection, giving optimised peak shapes
- Can be operated at pressures greater than 25000 psi
- SilFlow™ tracks oven temperature up to 20 °C/min. The design of SilFlow™ alleviates cold spots and sample condensation
: SilFlow™ kits contain: SilFlow™ microchannel device (MCD), SilTite™ FingerTite tool, mounting bracket, appropriately sized ferrules, nuts and blanking ferrules for set up. 1,1 mm Ø tubing is not supplied in the splitter kits and must be ordered separately.
Product Family Options
SilFlow™ GC 3-port splitters
Product Information
- AvailabilityPrice
- 548-1778GC kit (1,1), port A 1,1 mm OD, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm IDGC kit (1,1), port A 1,1 mm OD, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm IDLog in for availability1 * 1 ST826,00 €
- 548-1779GC kit (0,53), port A 0,53 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm IDGC kit (0,53), port A 0,53 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm IDLog in for availability1 * 1 ST826,00 €
- 548-1780GC kit (0,25/0,32), port A 0,25/0,32 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm IDGC kit (0,25/0,32), port A 0,25/0,32 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm IDLog in for availability1 * 1 ST826,00 €
- 548-1781GC MCD (1,1), port A 1,1 mm OD, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm IDGC MCD (1,1), port A 1,1 mm OD, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm IDLog in for availability1 * 1 ST543,00 €
- 548-1782GC MCD (0,53), port A 0,53 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm IDGC MCD (0,53), port A 0,53 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm IDLog in for availability1 * 1 ST543,00 €
- 548-1783GC MCD (0,25/0,32), port A 0,25/0,32 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm IDGC MCD (0,25/0,32), port A 0,25/0,32 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm IDLog in for availability1 * 1 ST543,00 €
SilFlow™ GC 4-port splitters
Product Information
- AvailabilityPrice
- 548-1785GC kit (1,1), port A 1,1 mm OD, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm ID, port D 0,25/0,32 mm IDGC kit (1,1), port A 1,1 mm OD, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm ID, port D 0,25/0,32 mm IDLog in for availability1 * 1 ST826,00 €
- 548-1786GC kit (0,53), port A 0,53 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm ID, port D 0,25/0,32 mm IDGC kit (0,53), port A 0,53 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm ID, port D 0,25/0,32 mm IDLog in for availability1 * 1 ST826,00 €
- 548-1787GC kit (0,25/0,32), port A 0,25/0,32 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm ID, port D 0,25/0,32 mm IDGC kit (0,25/0,32), port A 0,25/0,32 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm ID, port D 0,25/0,32 mm IDLog in for availability1 * 1 ST826,00 €
- 548-1788GC MCD (1,1), port A 1,1 mm OD, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm ID, port D 0,25/0,32 mm IDGC MCD (1,1), port A 1,1 mm OD, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm ID, port D 0,25/0,32 mm IDLog in for availability1 * 1 ST543,00 €
- 548-1789GC MCD (0,53), port A 0,53 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm ID, port D 0,25/0,32 mm IDGC MCD (0,53), port A 0,53 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm ID, port D 0,25/0,32 mm IDLog in for availability1 * 1 ST543,00 €
- 548-1790GC MCD (0,25/0,32), port A 0,25/0,32 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm ID, port D 0,25/0,32 mm IDGC MCD (0,25/0,32), port A 0,25/0,32 mm ID, port B 0,25/0,32 mm ID, port C 0,25/0,32 mm ID, port D 0,25/0,32 mm IDLog in for availability1 * 1 ST543,00 €
SilFlow™ GC Deans' switch
Product Information
SilFlow™ GC replacement parts
Product Information
- AvailabilityPrice
- 548-1771Ferrules 1,1 mm ID, for use with tubing of OD 1,07 mm
- 548-1800Ferrules 0,35 mm ID, for use with tubing of OD 0,32 mm
- 548-1772Ferrules 0,4 mm ID, for use with tubing of OD 0,36 mm
- 548-1773Ferrules 0,5 mm ID, for use with tubing of OD 0,45 mm
- 548-1774Ferrules 0,7 mm ID, for use with tubing of OD 0,68 mm
- 548-1802Ferrules 0,8 mm ID, for use with tubing of OD 0,79 mm
- 548-1770Nuts
- 548-1799LC nuts 1/32"
- 548-1801Blanking ferrules and pins
- 548-1777FingerTite tool
- SGEA123743Ferrules 0,55 mm ID
- SGEA123744Ferrules 0,75 mm ID
- SGEA123755Capillary tubing, stainless steel, OD×L 1,1×750 mm, sleeved to 1/16 " at one endCapillary tubing, stainless steel, OD×L 1,1×750 mm, sleeved to 1/16 " at one endLog in for availability1 * 1 ST46,80 €
SilFlow™ GC replacement fused silica tubing
Product Information
- AvailabilityPrice
- SGEA123750Deactivated silica tubing, length 2 m, 0,363 mm OD, 0,05 mm IDDeactivated silica tubing, length 2 m, 0,363 mm OD, 0,05 mm IDLog in for availability1 * 1 ST57,70 €
- SGEA123751Deactivated silica tubing, length 2 m, 0,363 mm OD, 0,1 mm IDDeactivated silica tubing, length 2 m, 0,363 mm OD, 0,1 mm IDLog in for availability1 * 1 ST57,70 €
- SGEA123752Deactivated silica tubing, length 2 m, 0,363 mm OD, 0,15 mm IDDeactivated silica tubing, length 2 m, 0,363 mm OD, 0,15 mm IDLog in for availability1 * 1 ST57,70 €
- SGEA123753Deactivated silica tubing, length 2 m, 0,363 mm OD, 0,2 mm IDDeactivated silica tubing, length 2 m, 0,363 mm OD, 0,2 mm IDLog in for availability1 * 1 ST57,70 €