ContactUsLinkComponent
Cleanroom gloves, nitrile, sterile, BioClean N-Plus™
Cleanroom gloves, nitrile, sterile, BioClean N-Plus™
Supplier:  Nitritex
Certificates
undefined
Cleanroom gloves, nitrile, sterile, BioClean N-Plus™
Supplier:  Nitritex
Certificates
Specifications
  • Catalog No:
  • 112-3869
  • 112-3871
  • 112-3867
  • 112-3866
  • 112-3868
  • 112-3865
  • 112-3872
  • 112-3870
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Cleanroom class:
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,10 mm (3,94 mil)
  • 0,10 mm (3,94 mil)
  • 0,10 mm (3,94 mil)
  • 0,10 mm (3,94 mil)
  • 0,10 mm (3,94 mil)
  • 0,10 mm (3,94 mil)
  • 0,10 mm (3,94 mil)
  • 0,10 mm (3,94 mil)
  • Finger thickness:
  • 0,21 mm (8.26 mil)
  • 0,21 mm (8.26 mil)
  • 0,21 mm (8.26 mil)
  • 0,21 mm (8.26 mil)
  • 0,21 mm (8.26 mil)
  • 0,21 mm (8.26 mil)
  • 0,21 mm (8.26 mil)
  • 0,21 mm (8.26 mil)
  • Palm thickness:
  • 0,18 mm (7,09 mil)
  • 0,18 mm (7,09 mil)
  • 0,18 mm (7,09 mil)
  • 0,18 mm (7,09 mil)
  • 0,18 mm (7,09 mil)
  • 0,18 mm (7,09 mil)
  • 0,18 mm (7,09 mil)
  • 0,18 mm (7,09 mil)
  • EN norm:
  • EN 455, EN 374 Type B
  • EN 455, EN 374 Type B
  • EN 455, EN 374 Type B
  • EN 455, EN 374 Type B
  • EN 455, EN 374 Type B
  • EN 455, EN 374 Type B
  • EN 455, EN 374 Type B
  • EN 455, EN 374 Type B