ContactUsLinkComponent
Dropping bulbs for pipettes and tubes
Dropping bulbs for pipettes and tubes
Catalog # KART3914
Supplier:  KARTELL
undefined
Dropping bulbs for pipettes and tubes
Catalog # KART3914
Supplier:  KARTELL
Specifications
  • Colour:
    Natural
  • Description:
    Dropping bulbs, height: 40 mm
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Dropping bulbs with rim, made of latex. Suitable for Ranvier dropping tubes and Pasteur pipettes.