ContactUsLinkComponent
Cleanroom gloves, latex, sterile, BioClean Maxima™
 :  Nitritex
undefined
Cleanroom gloves, latex, sterile, BioClean Maxima™
 :  Nitritex
  • Catalog No:
  • 112-3847
  • 112-3845
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Cleanroom class:
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • Cuff style:
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,12 mm
  • 0,12 mm
  • Finger thickness:
  • 0,22 mm
  • 0,22 mm
  • Palm thickness:
  • 0,20 mm
  • 0,20 mm
  • EN norm:
  • EN 455, EN 374
  • EN 455, EN 374
Frequently Bought Together