pH indicator papers, rolls, Whatman™
Universal indicator papers, roll, 1,0 - 11,0
N° de catalogue WHAT10362030
Fournisseur: Whatman products (Cytiva)
pH indicator papers, rolls, Whatman™
N° de catalogue WHAT10362030
Fournisseur: Whatman products (Cytiva)
Jump to:
Product Details & Documents
Universal indicator papers which have been impregnated with a mixture of several indicators. On contact with the sample solution they assume a particular colour. A check against the colour comparision table supplied allows the pH to be determined.
- Instant pH readings
- Accurate for a wide range of routine pH testing
- Convenient and portable for field use
Specifications
- pH range:1,0 - 11,0
- Description:Universal indicator papers, roll