Order Entry
Belgium
ContactUsLinkComponent
 
Steam indicator tape, Comply™
  489-0109
undefined
Steam indicator tape, Comply™
  489-0109

 

  • Width:
    12 mm
  • Description:
    Steam indicator tape, Comply™
  • Length:
    55 m
  • Seq:
    01
  • Pk:
    1 Roll

 

 

Indicator tapes are Class 1 process indicators used for exposure monitoring. They provide assurance to the operator handling the processed items that the pack has been exposed to the sterilisation process without the need to open the pack or check load control records. Optional tape dispensers are available for easy dispensing of indicator tapes.

  • Chemical process indicator changes colour when exposed to the steam sterilisation process to provide assurance that the packs were processed without the need to open the pack
  • Versatile tape adheres to all types of wraps
  • Allows the user to write on tapes