Order Entry
Belgium
ContactUsLinkComponent
VWR®, Bovine serum albumin (BSA)
VWR®, Bovine serum albumin (BSA)
Catalog # 421501J
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Bovine serum albumin (BSA)
Catalog # 421501J
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • CAS No.:
    9048-46-8
    90604-29-8
  • Source:
    Serum
  • Species:
    Bovine
  • Storage conditions:
    Refrigerator
  • Protein synonyms:
    BSA|Fraction V|Cohn Fraction V
  • Protein/peptide name:
    Bovine Serum Albumin
  • Purity:
    98%
  • Colour:
    off white to pale brown
  • pH:
    6.8-7.2
  • Grade:
    Standard Grade

Specifications

About this item

A standard grade albumin for basic applications, produced by a heat shock process in caprylic acid.

  • 5% Moisture