Orders LinkContactUsLinkComponent
Syringes
Syringes
Syringes
Certificates
2 Options Available Below
Product Details & Documents
These syringes are made from borosilicate glass. They are used as a replacement for pump syringes.
  • Accuracy and reproducibility of ±1%
  • Scale length: 60 mm
Thread in plunger button: 6–32 UNC.
Product Family Options
Product Information
  • Volume
    Type
    Availability
    Price
  • SGEA008787
    1/4-28 UNF termination
    1/4-28 UNF termination
    5 ml
    5MD-C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 167,00 €
    167,00 €
    1 * 1 ST
       167,00 €
  • SGEA008987
    1/4-28 UNF termination
    1/4-28 UNF termination
    10 ml
    10MD-C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 164,00 €
    164,00 €
    1 * 1 ST
       164,00 €