ContactUsLinkComponent
Syringes for HPLC autosamplers, Thermo Scientific instruments
Syringes for HPLC autosamplers, Thermo Scientific instruments
Catalog # 548-1644
undefined
Syringes for HPLC autosamplers, Thermo Scientific instruments
Catalog # 548-1644
Specifications
  • For:
    AS100/300 and AS1000/3000/3500 LC autosamplers
  • Type:
    250C-THERMOSPEC
  • Volume:
    250 µl
  • Description:
    Gas-tight, with PTFE-tipped plunger, 1/4-28 UNF front thread
  • Environmentally Preferable:
  • Pk:
    1