Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Adapters
Adapters, Joints with Tube Connectors
Connecting adapters, Clear-Seal®, WHEATON®
Connecting adapters, Clear-Seal®, WHEATON®
Connecting adapters, Clear-Seal®, WHEATON®
Catalog # WHEA139479
Supplier:
DWK Life Sciences
Expand image
Connecting adapters, Clear-Seal®, WHEATON®
Catalog # WHEA139479
Supplier:
DWK Life Sciences
Specifications
Socket size:
19/22 NS
Connector:
thread 13-425
Pk:
1
Specifications
About this item
Product Details
Product Details