Orders LinkContactUsLinkComponent
Multiple use earplugs, SmartFit®
Multiple use earplugs, SmartFit®
Supplier:   HONEYWELL SAFETY
Multiple use earplugs, SmartFit®
Supplier:  HONEYWELL SAFETY
Certificates
Multiple use earplugs, SmartFit®
Multiple use earplugs, SmartFit®
2 Options Available Below
Product Details & Documents
These earplugs with Conforming Material Technology™ use body heat to adapt to the shape of the ear canal for a comfortable, personalized fit.
  • Three-flange design delivers superior comfort and a truly individual fit
  • Detachable cord system
  • Metal ring on stem detectable by automated equipment (SmartFit Detectable only)
The detectable version with its bright blue color and metalized features can be detected by visual or automated methods and is therefore ideal for food applications.
SNR: 25 dB
Certifications: EN 352-2
Delivery information: Supplied with HearPack® storage case.
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • DALL1011239
    Multiple use earplugs with cord
    Multiple use earplugs with cord
    Blue
    Log in for availability
    Log in for availability
    1 * 50 PAIRE
    1 * 50 PAIRE - 90,60 €
    90,60 €
    1 * 50 PAIRE
       90,60 €
  • 111-0556
    Multiple use earplugs with cord, detectable
    Multiple use earplugs with cord, detectable
    Blue
    Log in for availability
    Log in for availability
    1 * 50 PAIRE
    1 * 50 PAIRE - 132,00 €
    132,00 €
    1 * 50 PAIRE
       132,00 €