Orders LinkContactUsLinkComponent
Multiple use earplugs, Quiet®
Multiple use earplugs, Quiet®
Earplugs Quiet, bell shape, without cord, Orange
Catalog # DALL1028456
Supplier:   HONEYWELL SAFETY
Multiple use earplugs, Quiet®
Catalog # DALL1028456
Supplier:  HONEYWELL SAFETY
Multiple use earplugs, Quiet®
Multiple use earplugs, Quiet®
Earplugs Quiet, bell shape, without cord, Orange
Price
52,60 €
Log in for availability
1 * 50 PAIRE
Product Details & Documents
These vinyl multiple earplugs are available with or without cord.
  • Bell shape: 'no-roll' design that is easy to handle and fit
  • Contoured shape comfortably matches contours of the ear canal
  • Smooth, non-irritating skin provides all-day comfort, easy to clean for long-term use
  • Built-in insertion stem makes insertion quick and easy
Suitable for use in chemical, manufacturing, medical, mining, municipal services, pharmaceutical and transportation fields.
SNR: 28 dB
NRR: 26
Certifications: EN 352-2
Specifications
  • Colour:
    Orange
  • Description:
    Earplugs Quiet, bell shape, without cord
  • Seq:
    01