ContactUsLinkComponent
Test strips, lactic acid, Reflectoquant®, Supelco®
Test strips, lactic acid, Reflectoquant®, Supelco®
Catalog # 1.16127.0001
Supplier:  Merck
CAS Number:  
Test strips, lactic acid, Reflectoquant®, Supelco®
Catalog # 1.16127.0001
Supplier:  Merck
CAS Number:  
Specifications
  • Chemical method:
    Enzymatic reaction
  • Pk:
    50 Tests
  • Description:
    Lactic acid test
  • Range:
    3 - 60,0 mg/l CH₃CHOHCO₂H
Specifications
About this item
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly
The test measures total lactic acid (sum of D- and L-lactic acid in beverages, e.g. wine, beer and fruit/vegetable juice after dilution. Furthermore the test can be used for the determination of lactic acid in food after pretreatment (typically dilution).