Orders LinkContactUsLinkComponent
Test strips, hydroxymethylfurfural (HMF), Reflectoquant®, Supelco®
Test strips, hydroxymethylfurfural (HMF), Reflectoquant®, Supelco®
Hydroxymethylfurfural (HMF) test, 1,0 - 60,0 mg/l (HMF), Enzymatic reaction, 50 Tests
Catalog # 1.17952.0001
Supplier:   Merck
Test strips, hydroxymethylfurfural (HMF), Reflectoquant®, Supelco®
Catalog # 1.17952.0001
Supplier:  Merck
CAS Number:  
Test strips, hydroxymethylfurfural (HMF), Reflectoquant®, Supelco®
Test strips, hydroxymethylfurfural (HMF), Reflectoquant®, Supelco®
Hydroxymethylfurfural (HMF) test, 1,0 - 60,0 mg/l (HMF), Enzymatic reaction, 50 Tests
CAS Number:  
Price
198,00 €
Log in for availability
1 * 50 Tests
Product Details & Documents
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly
Documents
Specifications
Specifications
  • Chemical method:
    Enzymatic reaction
  • Pk:
    50 Tests
  • Description:
    Hydroxymethylfurfural (HMF) test
  • Range:
    1,0 - 60,0 mg/l (HMF)
  • Cat. No.:
    1.17952.0001
  • M-Clarity quality level:
    MQ100
Specifications
Complementary Products
Product Information
  • Catalog #
    Description
    Model
    Availability
    Price
  • 1.17246.0001
    Reflectometer for test strips only
    RQflex 20
    Log in for availability
    1 * 1 ST - 1 420,00 €