Test strips, ascorbic acid, Reflectoquant®, Supelco®
Ascorbic acid test, 25 - 450 mg/l C₆H₈O₆, Phosphormolybdenum blue, 50 Tests
Supplier: Merck
Test strips, ascorbic acid, Reflectoquant®, Supelco®
Catalog # 1.16981.0001
Supplier: Merck
CAS Number:
Aller à :
Product Details & Documents
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly
Documents
Spécifications
Spécifications
- Chemical method:Phosphormolybdenum blue
- Pk:50 Tests
- Description:Ascorbic acid test
- Range:25 - 450 mg/l C₆H₈O₆
- Cat. No.:1.16981.0001
- M-Clarity quality level:MQ100
Spécifications
Complementary Products
Informations sur le produit
- Catalog #DescriptionModelAvailabilityPrice
- 1.17246.0001Reflectometer for test strips onlyRQflex 20Connexion Pour la disponibilité1 * 1 ST - 1 420,00 €