GC capillary column connectors, SilTite™ μ-unions
Fournisseur: Trajan Scientific and Medical
Certificats
À propos de cet article
Ferrules for 0,1 to 0,25 mm ID columns have 0,4 mm holes. Ferrules for 0,32 mm ID columns have 0,5 mm holes. Ferrules for 0,53 mm ID columns have 0,8 mm holes.
- Zero dead volume, ensuring optimised peak shapes
- Leak-free FingerTite technology
- Inert
- Stable for both extreme temperature and pressure cycling
- SilTite™ ferrules eliminate the need to re-tighten after installation
Livraison: Each pack is delivered with two unions, five double-taper ferrules and the necessary tools.
Frequently Bought Together