ContactUsLinkComponent
Reagent Kits for Protein and Fibre Analysis, Supelco®
Reagent Kits for Protein and Fibre Analysis, Supelco®
Catalog # 1.10307.0500
Supplier:  Merck
CAS Number:  
Reagent Kits for Protein and Fibre Analysis, Supelco®
Catalog # 1.10307.0500
Supplier:  Merck
CAS Number:  
Specifications
  • Chemical method:
    Biuret
  • Pk:
    500 mL
  • Description:
    Protein (Biuret Method at 546 nm), reagent solution for approx. 250 determinations
  • Environmentally Preferable:
  • M-Clarity quality level:
    MQ100
  • Tests per kit:
    ca.250