Orders LinkContactUsLinkComponent
Blank test strips, MQuant®, Supelco®
Blank test strips, MQuant®, Supelco®
Blank strip (for colour correction), MQ, 100 Tests
Catalog # 1.11860.0001
Supplier:   Merck
Blank test strips, MQuant®, Supelco®
Catalog # 1.11860.0001
Supplier:  Merck
CAS Number:  
Blank test strips, MQuant®, Supelco®
Blank test strips, MQuant®, Supelco®
Blank strip (for colour correction), MQ, 100 Tests
CAS Number:  
Price
34,20 €
Log in for availability
1 * 1 EA
Product Details & Documents
MQuant® (MQ) test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically-degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
  • Easy to handle: just dip, wait and measure visually against colour scale
  • Safe: no auxiliary reagents necessary
  • Immediate results within 2 minutes enables fast decisions
  • Portable: can be used inside or outside the laboratory
Documents
Specifications
Specifications
  • Pk:
    100 Tests
  • Description:
    Blank strip (for colour correction)
  • Type:
    MQ
  • Cat. No.:
    1.11860.0001
  • M-Clarity quality level:
    MQ100