Order Entry
Belgium
Orders LinkContactUsLinkComponent
QUANTOFIX® Test Strips, Calcium

QUANTOFIX® Test Strips, Calcium

QUANTOFIX® ascorbic acid, 0 • 50 • 100 • 200 • 300 • 500 • 700 • 1000 • 2000 - mg/L - Vitamin C; 25 - 1000 - mg/L - Vitamin C, Reduction method, 44×123×43 mm (packaging), 100 Tests, 100 Tests
Catalog # 705-0315
Supplier:   MACHEREY-NAGEL
QUANTOFIX® Test Strips, Calcium
Catalog # 705-0315
Supplier:  MACHEREY-NAGEL
CAS Number:  
QUANTOFIX® Test Strips, Calcium
QUANTOFIX® Test Strips, Calcium

QUANTOFIX® ascorbic acid, 0 • 50 • 100 • 200 • 300 • 500 • 700 • 1000 • 2000 - mg/L - Vitamin C; 25 - 1000 - mg/L - Vitamin C, Reduction method, 44×123×43 mm (packaging), 100 Tests, 100 Tests

CAS Number:  
Price
38,00 €
Log in for availability
1 * 1 EA
Product Details & Documents

QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. The test strips are intended for single use, maintenance or calibration are not required.

  • Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity

The test strips allow the quick and easy determination of calcium in solutions. They come with all necessary reagents so that the measurement can immediately be started. Within one minute reliable results are obtained. Calcium is an important element in nutrition and is therefore tested in foodstuff. It is essential for the building of human bones. Lack of calcium in the daily diet can lead to osteoporosis. The recommended consumption is about 1000 mg/day. In combination with magnesium, calcium is responsible for water hardness.

Specifications

Specifications

  • Chemical method:
    Reduction method
  • Pk:
    100 Tests
  • Description:
    QUANTOFIX® ascorbic acid
  • Dimensions:
    44×123×43 mm (packaging)
  • Tests per kit:
    100 Tests
  • Range:
    0 • 50 • 100 • 200 • 300 • 500 • 700 • 1000 • 2000 - mg/L - Vitamin C; 25 - 1000 - mg/L - Vitamin C
  • Cat. No.:
    705-0315
  • Parameters:
    All reagents included
  • Contents:
    100 test strips in a tube, instruction leaflet
  • Platform:
    Semi-quantitative test strips
  • Compliance:
    No hazardous material
  • Instrument:
    QUANTOFIX Relax
  • Shelf life (months):
    30 (from production)
  • Method:
    Ascorbic acid - Phosphomolybdenum blue (yellow → green-blue)
  • Weight:
    42 g

Specifications