Visual colour kits, pH indicator with colour card, Supelco®
Supplier: Merck
Certificates
About this item
Quick tests, visual colour comparison.
- Results in approximately 1 minute
- Easy to use
- Application-oriented measuring ranges
- Portable
Titrimetric and colorimetric methods.
This range comprises both titrimetric and colorimetric tests for medium concentrations. With its broad range of available test parameters, these quick tests are the method of choice for aquaria, fish farming, aquatic ecology, environmental, cooling and boiler water analysis.
Delivery: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together