Order Entry
Belgium
ContactUsLinkComponent
Visual colour kits, formaldehyde, MQuant®, Supelco®
Visual colour kits, formaldehyde, MQuant®, Supelco®
Catalog # 1.08028.0001
Supplier:  Merck
CAS Number:  
Visual colour kits, formaldehyde, MQuant®, Supelco®
Catalog # 1.08028.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Chemical method:
    Triazole
  • Pk:
    100 Tests
  • Description:
    Formaldehyde test
  • M-Clarity quality level:
    MQ100
  • Type:
    MTC
  • Range:
    0,1-0,25-0,4-0,6-0,8-1,0-1,5 mg/l HCHO

Specifications

About this item

Quick tests, visual colour comparison.

  • Results in approximately 5 minute
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available

This range comprises both titrimetric and colorimetric tests for medium concentrations. With its broad range of available test parameters, these quick tests are the method of choice for aquaria, fish farming, aquatic ecology, environmental, cooling and boiler water analysis.

Delivery: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.