Order Entry
Belgium
ContactUsLinkComponent
Lead(II) nitrate 0.1 mol/l (0.2 N) in aqueous solution Reag. Ph. Eur. 3003100
Lead(II) nitrate 0.1 mol/l (0.2 N) in aqueous solution Reag. Ph. Eur. 3003100
Catalog # REGE3003100
Supplier:  REAGECON
CAS Number:  
Lead(II) nitrate 0.1 mol/l (0.2 N) in aqueous solution Reag. Ph. Eur. 3003100
Catalog # REGE3003100
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :331.21 g/mol
EINECS:233-245-9
UN:3082
ADR:9,III
CAS Number:10099-74-8
Formula:Pb(NO₃)₂
MDL Number:MFCD00011153