Sodium hydroxide in aqueous solution in aqueous solution
Plastic container, 20 L
Supplier: VWR Collection
Sodium hydroxide in aqueous solution in aqueous solution
Catalog # VWRS6091.9020
Supplier: VWR Collection
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Density:1.003 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1824
- ADR:8,III
Specifications
Specifications
- Pack type:Plastic container
- Pk:20 L



