ContactUsLinkComponent
Burette and thermometer clamps with bosshead, AUTOMATIC
Burette and thermometer clamps with bosshead, AUTOMATIC
Catalog # BOCH5625
Supplier:  BOCHEM
undefined
Burette and thermometer clamps with bosshead, AUTOMATIC
Catalog # BOCH5625
Supplier:  BOCHEM
Specifications
  • For:
    Burettes and thermometers
  • Spanning width:
    0 - 25 mm
  • Pk:
    1
Specifications
About this item
These chromium plated clamps are designed for holding burettes or thermometers (Ø 0 - 25 mm). The bosshead (Ø 13,5 mm) can be rotated and locked at an angle of 90°.