9-Aminoacridine hydrochloride monohydrate ≥97.0% for synthesis, Sigma-Aldrich®
: Merck
: 9-Aminoacridine hydrochloride hydrate, Acridin-9-amine hydrochloride hydrate
:52417-22-8
:
:C₁₃H₁₁ClN₂·1H₂O
:MFCD00150071
:248.71 g/mol
:M-Clarity™ quality level = MQ200
:Ambient
:205-145-5
:2811
- Environmentally Preferable:
- CAS:52417-22-8
- Hill Formula:C₁₃H₁₁ClN₂ * H₂O
- Chemical formula:C₁₃H₁₀N₂ *HCl* H₂O
- Molar Mass:248.71 g/mol
- Melting Point:240 - 244 °C
- Solubility:3.3 g/l
- Assay (argentometric, calc. on anhydrous substance):≥ 97,0 %(m)
- Loss on drying:6,0 - 8,0 %(m)
- Identity (IR):conforms
Frequently Bought Together