Order Entry
Belgium
ContactUsLinkComponent
Mercury standard solution, 0,04 mg/L Hg in 10% nitric acid (from Hg(NO₃)₂)
Mercury standard solution, 0,04 mg/L Hg in 10% nitric acid (from Hg(NO₃)₂)
Catalog # CMPLD0BC.D04.10N.L
CAS Number:  
Mercury standard solution, 0,04 mg/L Hg in 10% nitric acid (from Hg(NO₃)₂)
Catalog # CMPLD0BC.D04.10N.L
CAS Number:  

Specifications

  • Concentration:
    0,04 mg/L
  • Matrix:
    10% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Mercury
  • Symbol:
    Hg

Specifications

About this item