Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 0.25 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Corrosive
Sodium hydroxide 0.25 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09958.0001
Supplier:  Merck
CAS Number:  
Corrosive
Sodium hydroxide 0.25 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09958.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • Seq:
    0501

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.185 g/cm3 (20 °C)
  • pH:
    14 (H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:∼105 °C (1013 hPa)
Density:1.327 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,
 :M-Clarity™ quality level = MQ100
EINECS:215-185-5
UN:1824
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548