
Sodium hydroxide 2% in aqueous solution, extra pure
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
:GHS05
Frequently Bought Together