Potassium thiocyanate 20% in aqueous solution, extra pure
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:333-20-0
:
Formula:KSCN
MDL Number:MFCD00011413
:97.18 g/mol
Storage Temperature:Ambient
EINECS:206-370-1
Frequently Bought Together