Order Entry
Belgium
Orders LinkContactUsLinkComponent
4-Phenylpyridine

4-Phenylpyridine

Glass bottle for solids, 25 g
Catalog # MOLE17588580-25G
Irritant
Marketsource
Supplier:   Molekula
4-Phenylpyridine
Catalog # MOLE17588580-25G
Supplier:  Molekula
Marketsource
CAS Number:  939-23-1
4-Phenylpyridine

Glass bottle for solids, 25 g

CAS Number:  939-23-1
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:939-23-1
  • Formula:C₁₁H₉N
  • MDL Number:MFCD00006420
  • Molecular Weight:155.2 g/mol
  • Boiling Pt:274 °C (1013 hPa)
  • Melting Pt:78 °C
  • Storage Temperature:Ambient
  • EINECS:213-357-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications