Order Entry
Belgium
ContactUsLinkComponent
Pyridine, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Flammable
Irritant
Pyridine, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Flammable
Irritant
Pyridine, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -42 °C
  • Molar Mass:
    79.1 g/mol
  • Boiling Point:
    115 °C (1013 hPa)
  • Vapor Pressure:
    20 hPa (20 °C)
  • Flash Point:
    17 °C
  • Explosion Limit:
    1.7 - 10.6 %(V)
  • Density:
    0.982 g/cm3 (20 °C)
  • pH:
    8.5 (16 g/l, H2O, 20 °C)
  • Ignition Point:
    482 °C
  • Evaporation number:
    12.7

Specification Test Results

Related Information

Frequently Bought Together